Ethyl-6-amino-2-phenyl-1H-indole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-amino-2-phenyl-1H-indole-3-carboxylate

072332-250MG 072332-1G
102524-296EA 366.15 USD
102524-296 102524-298
Ethyl-6-amino-2-phenyl-1H-indole-3-carboxylate ≥95%
Ethyl-6-amino-2-phenyl-1H-indole-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₆N₂O₂
MW: 280.32 g/mol
MDL Number: MFCD09953516
CAS Number: 945655-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR