4-Methoxy-2-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072331-1G 072331-5G
102524-290EA 73.85 USD
102524-290 102524-292
4-Methoxy-2-nitrobenzaldehyde ≥95%
4-Methoxy-2-nitrobenzaldehyde

Hazard: Irritant/Store in Freezer

Formula: C₈H₇NO₄
MW: 181.1455 g/mol
MDL Number: MFCD00024254
CAS Number: 22996-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR