2-Amino-6-methoxypyrazine ≥95%

Supplier: Matrix Scientific

072327-1G 072327-5G
102524-268EA 516.92 USD
102524-268 102524-270
2-Amino-6-methoxypyrazine ≥95%
2-Amino-6-methoxypyrazine

Hazard: Irritant

Formula: C₅H₇N₃O
MW: 125.13 g/mol
MDL Number: MFCD09909689
CAS Number: 6905-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR