3-Amino-5-(4-chlorophenyl)thiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

072326-250MG 072326-1G 072326-5G
102524-262EA 400 USD
102524-262 102524-264 102524-266
3-Amino-5-(4-chlorophenyl)thiophene-2-carboxylic acid ≥95%
3-Amino-5-(4-chlorophenyl)thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C11H8ClNO2S
MW: 253.71 g/mol
MDL Number: MFCD09909681
CAS Number: 187949-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR