2,4-Dibromothiazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

072325-250MG 072325-1G
102524-258EA 480 USD
102524-258 102524-260
2,4-Dibromothiazole-5-carboxylic acid ≥95%
2,4-Dibromothiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₄HBr₂NO₂S
CAS Number: 139669-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR