2,4-Dichlorothiazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

072323-250MG 072323-1G 072323-5G
102524-248EA 121.54 USD
102524-248 102524-250 102524-252
2,4-Dichlorothiazole-5-carboxylic acid ≥95%
2,4-Dichlorothiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₄HCl₂NO₂S
MW: 198.0300 g/mol
MDL Number: MFCD09909676
CAS Number: 62019-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR