5-Chloropyrido[3,4-b]pyrazine ≥95%

Supplier: Matrix Scientific

072322-250MG 072322-1G
102524-244EA 235.38 USD
102524-244 102524-246
5-Chloropyrido[3,4-b]pyrazine ≥95%
5-Chloropyrido[3,4-b]pyrazine

Hazard: Irritant

Formula: C7H4ClN3
MW: 165.5800 g/mol
MDL Number: MFCD08706209
CAS Number: 214045-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR