Ethyl [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1,2,4-Triazolo[4,3-a]pyrazine-3-carboxylate

072321-250MG 072321-1G
102524-240EA 498.46 USD
102524-240 102524-242
Ethyl [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate ≥95%
Ethyl [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate

Hazard: Irritant

Formula: C₈H₈N₄O₂
MW: 192.18 g/mol
MDL Number: MFCD09909674
CAS Number: 723286-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR