2-Fluoro-5-hydroxybenzaldehyde ≥95%
Supplier: Matrix Scientific
072319-1G
072319-5G
102524-228EA
89.23
USD
102524-228
102524-230
2-Fluoro-5-hydroxybenzaldehyde ≥95%
2-Fluoro-5-hydroxybenzaldehyde
Hazard: Irritant
Formula:
C₇H₅FO₂ MW: 140.1100 g/mol |
MDL Number:
MFCD09038469 CAS Number: 103438-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...