6-Hydroxybenzo[d]thiazole-2-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-hydroxybenzo[d]thiazole-2-carbonitrile, 2-Cyano-6-hydroxybenzothiazole, 6-hydroxy-1,3-benzothiazole-2-carbonitrile

072318-250MG 072318-1G 072318-5G
102524-222EA 140 USD
102524-222 102524-224 102524-226
6-Hydroxybenzo[d]thiazole-2-carbonitrile ≥95%
6-Hydroxybenzo[d]thiazole-2-carbonitrile

Hazard: Irritant

Formula: C₈H₄N₂OS
MDL Number: MFCD00296905
CAS Number: 939-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR