3-Oxo-3,4-dihydroquinoxaline-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-quinoxalinecarboxylic acid

072315-250MG 072315-1G 072315-5G
102524-204EA 498.46 USD
102524-204 102524-206 102524-208
3-Oxo-3,4-dihydroquinoxaline-2-carboxylic acid ≥95%
3-Oxo-3,4-dihydroquinoxaline-2-carboxylic acid

Hazard: Irritant

Formula: C9H6N2O3
CAS Number: 1204-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR