3-Bromo-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one ≥95%
Supplier: Matrix Scientific
072311-1G
072311-5G
102524-186EA
200
USD
102524-186
102524-188
3-Bromo-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one ≥95%
3-Bromo-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
Hazard: Irritant
Formula:
C5H3BrN4O MW: 215.0100 g/mol |
MDL Number:
MFCD00835293 CAS Number: 54738-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...