2-Hydroxy-5-nitropyrimidine ≥95%
Supplier: Matrix Scientific
072308-1G
072308-5G
102524-174EA
267.69
USD
102524-174
102524-176
2-Hydroxy-5-nitropyrimidine ≥95%
2-Hydroxy-5-nitropyrimidine
Hazard: Irritant
Formula:
C₄H₃N₃O₃ Melting Pt: 203 °C |
CAS Number:
3264-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...