2-(4-Bromophenyl)-1H-imidazo[4,5-c]pyridine ≥95%

Supplier: Matrix Scientific

072305-250MG 072305-1G
102524-160EA 967.69 USD
102524-160 102524-162
2-(4-Bromophenyl)-1H-imidazo[4,5-c]pyridine ≥95%
2-(4-Bromophenyl)-1H-imidazo[4,5-c]pyridine

Hazard: Irritant

Formula: C₁₂H₈BrN₃
MW: 274.1200 g/mol
MDL Number: MFCD06657714
CAS Number: 113270-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR