2-Amino-6-bromopyrido[2,3-d]pyrimidin-4(3H)-one ≥95%
Supplier: Matrix Scientific
072304-250MG
072304-1G
102524-156EA
640
USD
102524-156
102524-158
2-Amino-6-bromopyrido[2,3-d]pyrimidin-4(3H)-one ≥95%
2-Amino-6-bromopyrido[2,3-d]pyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₇H₅BrN₄O |
CAS Number:
120040-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...