7-Bromopyrido[2,3-b]pyrazine-2,3(1H,4H)-dione ≥95%

Supplier: Matrix Scientific

072303-250MG 072303-1G 072303-5G
102524-150EA 640 USD
102524-150 102524-152 102524-154
7-Bromopyrido[2,3-b]pyrazine-2,3(1H,4H)-dione ≥95%
7-Bromopyrido[2,3-b]pyrazine-2,3(1H,4H)-dione

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
MW: 242.0300 g/mol
MDL Number: MFCD00460131
CAS Number: 168123-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR