Thiazolo[5,4-c]pyridin-2-amine ≥95%

Supplier: Matrix Scientific

072302-250MG 072302-1G
102524-146EA 967.69 USD
102524-146 102524-148
Thiazolo[5,4-c]pyridin-2-amine ≥95%
Thiazolo[5,4-c]pyridin-2-amine

Hazard: Irritant

Formula: C₆H₅N₃S
MW: 151.190 g/mol
MDL Number: MFCD09909658
CAS Number: 108310-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR