2-(Azetidin-1-yl)-5-bromopyrimidine ≥95%

Supplier: Matrix Scientific

072301-250MG 072301-1G 072301-5G
102524-140EA 498.46 USD
102524-140 102524-142 102524-144
2-(Azetidin-1-yl)-5-bromopyrimidine ≥95%
2-(Azetidin-1-yl)-5-bromopyrimidine

Hazard: Irritant

Formula: C₇H₈BrN₃
MW: 214.0600 g/mol
MDL Number: MFCD08706083
CAS Number: 850349-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR