3-Bromo-8-chloroimidazo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

072298-1G 072298-5G
102524-124EA 201.54 USD
102524-124 102524-126
3-Bromo-8-chloroimidazo[1,2-a]pyrazine ≥95%
3-Bromo-8-chloroimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C6H3BrClN3
MDL Number: MFCD09909653
CAS Number: 143591-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR