2-Bromothiazole-5-carbonitrile ≥95%

Supplier: Matrix Scientific

072296-1G 072296-5G
102524-114EA 710.77 USD
102524-114 102524-116
2-Bromothiazole-5-carbonitrile ≥95%
2-Bromothiazole-5-carbonitrile

Hazard: Irritant

Formula: C₄HBrN₂S
MW: 189.0300 g/mol
MDL Number: MFCD09909651
CAS Number: 440100-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR