6-Bromo-1,8-naphthyridin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-bromo-1,8-naphthyridine

072292-250MG 072292-1G
102524-098EA 967.69 USD
102524-098 102524-100
6-Bromo-1,8-naphthyridin-2-amine ≥95%
6-Bromo-1,8-naphthyridin-2-amine

Hazard: Irritant

Formula: C₈H₆BrN₃
MW: 224.0573 g/mol
MDL Number: MFCD09909648
CAS Number: 64874-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR