2-Amino-5-methoxypyrazine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Methoxypyrazin-2-amine
072291-1G
072291-5G
102524-094EA
283.08
USD
102524-094
102524-096
2-Amino-5-methoxypyrazine ≥95%
2-Amino-5-methoxypyrazine
Hazard: Irritant
Formula:
C₅H₇N₃O MW: 125.13 g/mol |
MDL Number:
MFCD09909647 CAS Number: 54013-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...