Ethyl-4-chloropyrimidine-5-carboxylate ≥95%

Supplier: Matrix Scientific

072288-1G 072288-5G 072288-10G
102524-080EA 135.38 USD
102524-080 102524-082 102524-084
Ethyl-4-chloropyrimidine-5-carboxylate ≥95%
Ethyl-4-chloropyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₇H₇ClN₂O₂
MW: 186.6 g/mol
MDL Number: MFCD07371659
CAS Number: 41103-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR