Isoquinoline-4-carbonitrile ≥95%

Supplier: Matrix Scientific

072287-1G 072287-5G
102524-076EA 710.77 USD
102524-076 102524-078
Isoquinoline-4-carbonitrile ≥95%
Isoquinoline-4-carbonitrile

Hazard: Irritant

Formula: C₁₀H₆N₂
MW: 154.1700 g/mol
MDL Number: MFCD00957039
CAS Number: 34846-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR