3-Chloropyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

072286-1G 072286-5G
102524-070EA 44.62 USD
102524-070 102524-072
3-Chloropyrazine-2-carboxylic acid ≥95%
3-Chloropyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C5H3ClN2O2
Melting Pt: 116…117 °C
MDL Number: MFCD01318438
CAS Number: 27398-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR