1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

072284-1G 072284-5G
102524-062EA 710.77 USD
102524-062 102524-064
1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine ≥95%
1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₈H₁₂N₂
MDL Number: MFCD00296984
CAS Number: 73627-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR