3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride ≥95%

Supplier: Matrix Scientific

072281-250MG 072281-1G
102524-048EA 748.05 USD
102524-048 102524-050
3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride ≥95%
3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride

Hazard: Irritant

Formula: C7H16Cl2N2
MDL Number: MFCD18083726
CAS Number: 52407-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR