(5-Fluoro-2-nitrophenyl)acetic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Fluoro-2-nitrophenylacetic acid
072277-1G
072277-5G
072277-10G
102524-024EA
233.85
USD
102524-024
102524-026
102524-028
(5-Fluoro-2-nitrophenyl)acetic acid ≥95%
(5-Fluoro-2-nitrophenyl)acetic acid
Hazard: Irritant
Formula:
C₈H₆FNO₄ MW: 199.14 g/mol |
MDL Number:
MFCD07787601 CAS Number: 29640-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...