N-Boc-Piperidin-4-ylacetic acid methyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Boc-Piperidin-4-yl-acetic acid methyl ester, Methyl (piperidin-4-yl)acetate, N-BOC protected
072276-1G
072276-5G
102524-020EA
267.69
USD
102524-020
102524-022
N-Boc-Piperidin-4-ylacetic acid methyl ester ≥95%
N-Boc-Piperidin-4-ylacetic acid methyl ester
Hazard: Irritant
Formula:
C₁₃H₂₃NO₄ MW: 257.3300 g/mol |
MDL Number:
MFCD01632529 CAS Number: 175213-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...