5-Methoxy-2-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072275-5G 072275-25G
102524-014EA 138.46 USD
102524-014 102833-530
5-Methoxy-2-nitrobenzaldehyde ≥95%
5-Methoxy-2-nitrobenzaldehyde

Hazard: Irritant

Formula: C₈H₇NO₄
Melting Pt: 80-82°C
MDL Number: MFCD00456498
CAS Number: 20357-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR