Ethyl-5-aminopyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-aminopyrazine-2-carboxylate

072274-1G 072274-5G
102524-008EA 123.08 USD
102524-008 102524-010
Ethyl-5-aminopyrazine-2-carboxylate ≥95%
Ethyl-5-aminopyrazine-2-carboxylate

Hazard: Irritant

Formula: C₇H₉N₃O₂
MW: 167.17 g/mol
MDL Number: MFCD08437665
CAS Number: 54013-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR