N-Boc-3-ethylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Ethylpiperazine, N1-BOC protected, N-Boc-3-Ethylpiperazine

072270-1G 072270-5G
102523-988EA 533.85 USD
102523-988 102523-990
N-Boc-3-ethylpiperazine ≥95%
N-Boc-3-ethylpiperazine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
MDL Number: MFCD07367735
CAS Number: 438049-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR