2-Hydroxypyridine-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxyisonicotinaldehyde, 2-Hydroxy-4-formylpyridine

072269-1G 072269-5G
102523-984EA 435.38 USD
102523-984 102523-986
2-Hydroxypyridine-4-carbaldehyde ≥95%
2-Hydroxypyridine-4-carbaldehyde

Hazard: Irritant

Formula: C₆H₅NO₂
MW: 123.11 g/mol
MDL Number: MFCD03788455
CAS Number: 188554-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR