2,4-Bis(benzyloxy)-5-bromopyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2,4-di(benzyloxy)pyrimidine

072266-1G 072266-5G 072266-10G
102523-968EA 213.85 USD
102523-968 102523-970 102523-972
2,4-Bis(benzyloxy)-5-bromopyrimidine ≥95%
2,4-Bis(benzyloxy)-5-bromopyrimidine

Hazard: Irritant

Formula: C₁₈H₁₅BrN₂O₂
MW: 371.2300 g/mol
MDL Number: MFCD00267932
CAS Number: 41244-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR