5-Bromo-4,6-dihydroxypyrimidine ≥95%

Supplier: Matrix Scientific

072264-1G 072264-5G
102523-958EA 249.23 USD
102523-958 102523-960
5-Bromo-4,6-dihydroxypyrimidine ≥95%
5-Bromo-4,6-dihydroxypyrimidine

Hazard: Irritant

Formula: C₄H₃BrN₂O₂
MW: 190.9828 g/mol
MDL Number: MFCD00047380
CAS Number: 15726-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR