2-Amino-4,6-dichloro-5-methylpyrimidine ≥95%

Supplier: Matrix Scientific

072263-1G 072263-5G
102523-954EA 521.54 USD
102523-954 102523-956
2-Amino-4,6-dichloro-5-methylpyrimidine ≥95%
2-Amino-4,6-dichloro-5-methylpyrimidine

Hazard: Irritant

Formula: C₅H₅Cl₂N₃
MDL Number: MFCD00023253
CAS Number: 7153-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR