Ethyl-6-chloro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

072261-250MG 072261-1G 072261-5G
102523-946EA 498.46 USD
102523-946 102523-948 102551-792
Ethyl-6-chloro-3-pyridazinecarboxylate ≥95%
Ethyl-6-chloro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₇H₇ClN₂O₂
MDL Number: MFCD09908168
CAS Number: 75680-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR