1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
7-Amino-N-methyl-1,2,3,4-tetrahydroquinoline hydrochloride
072260-250MG
072260-1G
072260-5G
102523-940EA
307.69
USD
102523-940
102523-942
102523-944
1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride ≥95%
1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂ MW: 198.69 g/mol |
MDL Number:
MFCD09839668 CAS Number: 927684-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...