4-Benzyl-2-morpholinecarboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Benzylmorpholine-2-carboxylic acid hydrochloride

072259-1G 072259-5G 072259-10G
102523-934EA 284.62 USD
102523-934 102523-936 102523-938
4-Benzyl-2-morpholinecarboxylic acid hydrochloride ≥95%
4-Benzyl-2-morpholinecarboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₃
MW: 257.71 g/mol
MDL Number: MFCD02682031
CAS Number: 135072-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR