4,5,6,7-Tetrahydro-2-methylthiazolo[5,4-c]pyridine ≥95%

Supplier: Matrix Scientific

072254-250MG 072254-1G 072254-5G
102523-906EA 429.23 USD
102523-906 102523-908 102523-910
4,5,6,7-Tetrahydro-2-methylthiazolo[5,4-c]pyridine ≥95%
4,5,6,7-Tetrahydro-2-methylthiazolo[5,4-c]pyridine

Hazard: Irritant

Formula: C₇H₁₀N₂S
MW: 154.2327 g/mol
MDL Number: MFCD06738531
CAS Number: 124458-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR