5-Fluoro-1H-indazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

072253-1G 072253-5G
102523-902EA 640 USD
102523-902 102523-904
5-Fluoro-1H-indazole-3-carboxylic acid ≥95%
5-Fluoro-1H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₅FN₂O₂
MW: 180.140 g/mol
MDL Number: MFCD03840628
CAS Number: 1077-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR