6-Chloroimidazo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

072252-1G 072252-5G
102523-898EA 258.46 USD
102523-898 102523-900
6-Chloroimidazo[1,2-a]pyrazine ≥95%
6-Chloroimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
MDL Number: MFCD09909644
CAS Number: 76537-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR