3-Bromo-7-(trifluoromethyl)imidazo[1,2-a]pyrimidine ≥95%

Supplier: Matrix Scientific

072251-250MG 072251-1G 072251-5G
102523-892EA 364.62 USD
102523-892 102523-894 102523-896
3-Bromo-7-(trifluoromethyl)imidazo[1,2-a]pyrimidine ≥95%
3-Bromo-7-(trifluoromethyl)imidazo[1,2-a]pyrimidine

Hazard: Irritant

Formula: C7H3BrF3N3
MW: 266.0180 g/mol
MDL Number: MFCD08234995
CAS Number: 375857-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR