2-Bromo-5-hydroxypyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromopyrazin-2-ol

072250-5G 072250-25G
102523-888EA 106.15 USD
102523-888 102663-872
2-Bromo-5-hydroxypyrazine ≥95%
2-Bromo-5-hydroxypyrazine

Hazard: Irritant

Formula: C₄H₃BrN₂O
MW: 174.9800 g/mol
MDL Number: MFCD06245330
CAS Number: 374063-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR