2-Amino-5-bromo-3-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-3-hydroxypyridine

072249-1G 072249-5G
102523-882EA 23.08 USD
102523-882 102523-884
2-Amino-5-bromo-3-pyridinol ≥95%
2-Amino-5-bromo-3-pyridinol

Hazard: Irritant

Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
MDL Number: MFCD09744143
CAS Number: 39903-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR