7-Aminoquinoline ≥95%

Supplier: Matrix Scientific

072248-1G 072248-5G
102523-878EA 136.92 USD
102523-878 102523-880
7-Aminoquinoline ≥95%
7-Aminoquinoline

Hazard: Irritant

Formula: C₉H₈N₂
MW: 144.18 g/mol
MDL Number: MFCD01646243
CAS Number: 580-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR