7-Nitroquinoline ≥95%
Supplier: Matrix Scientific
072247-1G
072247-5G
102523-872EA
640
USD
102523-872
102523-874
7-Nitroquinoline ≥95%
7-Nitroquinoline
Hazard: Irritant
Formula:
C₉H₆N₂O₂ MW: 174.1561 g/mol |
MDL Number:
MFCD00234498 CAS Number: 613-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...