5-Chloropyrimidin-2(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-hydroxy-pyrimidine·HCl

072246-1G 072246-5G
102523-868EA 427.69 USD
102523-868 102523-870
5-Chloropyrimidin-2(1H)-one ≥95%
5-Chloropyrimidin-2(1H)-one

Hazard: Irritant

Formula: C4H3ClN2O
MDL Number: MFCD09250871
CAS Number: 54326-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR