2-Amino-5-bromo-6-chloropyrazine ≥95%

Supplier: Matrix Scientific

072244-1G 072244-5G 072244-10G
102523-858EA 340 USD
102523-858 102523-860 102523-862
2-Amino-5-bromo-6-chloropyrazine ≥95%
2-Amino-5-bromo-6-chloropyrazine

Hazard: Irritant

Formula: C₄H₃BrClN₃
MDL Number: MFCD09909642
CAS Number: 173253-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR