Methyl-3-amino-6-iodopyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-6-iodopyrazine-2-carboxylate

072243-250MG 072243-1G
102523-854EA 541.54 USD
102523-854 102523-856
Methyl-3-amino-6-iodopyrazine-2-carboxylate ≥95%
Methyl-3-amino-6-iodopyrazine-2-carboxylate

Hazard: Irritant

Formula: C₆H₆IN₃O₂
MW: 279.04 g/mol
MDL Number: MFCD09909640
CAS Number: 1458-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR